Coliform Sample Container
Actual product may vary
Corning
Coliform Sample Container
Automatically dechlorinates sample
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
FIS09-730-91Aw/o Sodium Thiosulfate Tablet
CRG1705-100w/o Sodium Thiosulfate Tablet
CRG1700-100Sodium Thiosulfate Tablet
Catalog number FIS09-730-91A
Price (MYR)
796.00
Case
Includes:
w/o Sodium Thiosulfate Tablet
Price (MYR)
796.00
Case
These Corning™ containers are used to test nonchlorinated drinking water.
  • Manufactured from pure, recyclable polypropylene.
  • Capacity: 120 mL total capacity.
  • Capacity at line: 100 mL ± 2.5 mL (2.5%).
  • The one-piece container has attached lid to reduce chance of contamination.
  • Locking arrow assures sterility has not been compromised.
  • Leak tight when sealed properly. An added benefit is the tie-down which protects against accidental opening.
  • Sterile-by-process
Specifications
Certifications/ComplianceEPA
Closure ColorNatural
Closure MaterialPolypropylene
Closure TypeSnap Cap
Volume (Metric)120 mL
DisposableSingle-use
For Use With (Application)Test for the presence of coliform bacteria, microbiologic contaminants in drinking water
GraduatedYes
Includesw/o Sodium Thiosulfate Tablet
Neck StyleWide Mouth
SterilitySterile
Volume (English)4 oz.
Height (English)3.77 in.
Height (Metric)9.6 cm
Diameter (English) Outer1.82 in.
Diameter (Metric) Outer4.64 cm
Unit SizeCase