Heavy Glass Desiccators
Heavy Glass Desiccators
Actual product may vary
Fisherbrand
Heavy Glass Desiccators
Annealed glass provides durability
Have Questions?
Catalog NumberDiameter (Metric) PlateDiameter (Metric) Inner
FIS08-595E23 cm25 cm
Catalog number FIS08-595E
Price (MYR)
1,749.00
EA
Add to cart
Diameter (Metric) Plate:
23 cm
Diameter (Metric) Inner:
25 cm
Price (MYR)
1,749.00
EA
Add to cart
Annealed glass provides durability
  • Apply grease to internal edges of flanges
  • Press lid on jar and rotate at least 360° to ensure airtight seal
  • Rim in lower portion of jar holds sample plate over desiccant
Specifications
MaterialAnnealed Glass
Product TypeNonvacuum Desiccator
IncludesCover
Diameter (Metric) Inner25 cm
Diameter (Metric) Plate23 cm
Unit SizeEA