▸
Organic compounds
▸
Unclassified Organic Compounds
▸
Kjeldahl Selenium Catalyst Tablets, Technical, Contains 1g Na2SO4 And Eq. Of 0.05g Selenium, Fisher Chemical™
Element Search
Structure Search
Kjeldahl Selenium Catalyst Tablets, Technical, Contains 1g Na2SO4 And Eq. Of 0.05g Selenium, Fisher Chemical™
Kjeldahl Selenium Catalyst Tablets, Technical, Contains 1g Na2SO4 And Eq. Of 0.05g Selenium, Fisher Chemical, Quantity: 1kg, Gray, MDL Number: 147338, Packaging: Plastic powder jar, Physical Form: Solid | CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change view
Quantity:
1 kg
0.5 kg
Catalog number
FGCK015081
also known as FGCK/0150/81
Price (MYR)
1,598.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,598.00
Each
Add to cart
Specifications
CAS
7782-49-2
Color
Gray
Packaging
Plastic powder jar
Molecular Formula
Se
MDL Number
MFCD00134090 MFCD00011224
View more
Kjeldahl Selenium Catalyst Tablets, Technical, Contains 1g Na2SO4 And Eq. Of 0.05g Selenium, Fisher Chemical, Quantity: 1kg, Gray, MDL Number: 147338, Packaging: Plastic powder jar, Physical Form: Solid