▸
Bases
▸
Sodium Hydroxide
▸
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Have Questions?
Change view
Quantity:
5 L
2.5 L
10 L
Packaging:
View all
Plastic container
Catalog number
FGCJ762021
also known as FGCJ/7620/21
Price (MYR)
423.00
Each
Add to cart
Quantity:
5 L
Packaging:
Plastic container
Request bulk or custom format
Price (MYR)
423.00
Each
Add to cart
Specifications
Color
Colorless
Boiling Point
100°C
CAS
1310-73-2
Molecular Formula
HNaO
MDL Number
3548
View more
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp