Chloramphenicol (Crystalline Powder), Fisher BioReagents
Chloramphenicol (Crystalline Powder), Fisher BioReagents
Chloramphenicol (Crystalline Powder), Fisher BioReagents
Fisher Chemical
Chloramphenicol (Crystalline Powder), Fisher BioReagents
For selecting transformed cells containing chloramphenicol resistance genes
Have Questions?
Catalog NumberQuantity
FGCBP904-100100 g
Catalog number FGCBP904-100
Price (MYR)
948.00
Each
Quantity:
100 g
Request bulk or custom format
Price (MYR)
948.00
Each
Specifications
Chemical Name or MaterialChloramphenicol
TypeChloramphenicol
Physical FormSolid
CAS56-75-7
ChemAlert Storage SymbolGray
View more
Used as a selection agent for transformed cells containing chloramphenicol resistance genes. Mode of Action: Inhibits translation on the 50S ribosomal subunit at the peptidyltransferase step (elongation inhibition).