▸
Buffers and Standards
▸
Life Science Buffers
▸
MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Commonly used buffering agent
Have Questions?
Change view
Quantity:
500 g
100 g
Packaging:
View all
Poly Bottle
Catalog number
FGCBP308-500
Price (MYR)
934.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
934.00
Each
Add to cart
Specifications
Chemical Name or Material
MOPS
CAS
1132-61-2
ChemAlert Storage Symbol
Gray
Melting Point
277°C
Boiling Point
277°C
View more
Commonly used buffering agent
This biological buffer has a usable pH range of 6.5 to 7.9.