▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Reagents, Antibiotics and Supplements
▸
Antibiotics
▸
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
For selection for ampicillin resistance in mutated and transformed cells | CAS: 69-52-3 | C16H21N3NaO4S | 374.411 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FGCBP1760-5
5 g
Poly Bottle
Catalog number
FGCBP1760-5
Price (MYR)
642.00
Each
Add to cart
Quantity:
5 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
642.00
Each
Add to cart
Specifications
Chemical Name or Material
Ampicillin Sodium Salt
Type
Ampicillin Sodium Salt
Physical Form
Solid
CAS
69-52-3
ChemAlert Storage Symbol
Blue
View more
For selection for ampicillin resistance in mutated and transformed cells
For selection for ampicillin resistance in mutated and transformed cells. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.