Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™
Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™
Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™
Fisher Chemical
Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™
Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number FGCBP172-5
Price (MYR)
629.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
629.00
Each
Specifications
Chemical Name or MaterialDithiothreitol
CAS12-3-3483
ChemAlert Storage SymbolGray
Absorbance0.04 AU max.
Melting Point38°C
View more
Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %
Dithiothreitol is used to reduce disulfide bonds in proteins and peptides and is added in low concentrations as an enzyme stabilizer during protein purification and enzymatic reactions.