Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
Fisher Chemical
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
For electrophoresis, molecular biology, and cell culture applications | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Catalog number FGCBP153-500
Price (MYR)
649.00
Each
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
649.00
Each
Specifications
Absorbance0.05 max. (1M solution) at 280nm
Chemical Name or MaterialTris Hydrochloride
CAS1185-53-1
ChemAlert Storage SymbolGray
Melting Point150°C
View more
For electrophoresis, molecular biology, and cell culture applications
Tris Hydrochloride is a buffering medium for electrophoresis, molecular biology, and cell culture applications. It may be combined with Tris Base to simplify Tris buffer preparation.