TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™
TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™
TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™
TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™
Fisher Chemical
TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™
Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Packaging:
View all
Nalgene Poly Bottle
Catalog number FGCBP1332-1
Price (MYR)
552.00
EA
Add to cart
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Request bulk or custom format
Price (MYR)
552.00
EA
Add to cart
Chemical Identifiers
CAS77-86-1, 60-00-4, 6850-28-8
Specifications
Chemical Name or MaterialTris-Acetate-EDTA
Name Note50X Solution
ChemAlert Storage SymbolGray
CAS77-86-1
Recommended StorageRT
View more
Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT
Tris–Acetate–EDTA (TAE) is commonly used as a buffer for nucleic acid electrophoresis.