▸
Bases
▸
Ammonium Hydroxide
▸
Ammonia Solution 20-22%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Ammonia Solution 20-22%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCA512-P500
500 mL
Catalog number
FGCA512-P500
Price (MYR)
686.00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
686.00
Each
Add to cart
Specifications
pH
13.6
Color
Colorless
CAS
1336-21-6
Molecular Formula
H
5
NO
MDL Number
66650
View more