Methanol, Optima™ LC/MS Grade, Fisher Chemical
Methanol, Optima™ LC/MS Grade, Fisher Chemical
Methanol, Optima™ LC/MS Grade, Fisher Chemical
Fisher Chemical
Methanol, Optima™ LC/MS Grade, Fisher Chemical
Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol | CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantity
FGCA456-2122.5 L
Catalog number FGCA456-212
Price (MYR)
751.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
751.00
Each
Specifications
Chemical Name or MaterialMethanol
Melting Point-98°C
Absorbance≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.2 AU at 220nm, ≤0.4 AU at 214nm, ≤0.5 AU at 210nm
CAS67-56-1
ColorColorless
View more
Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol
Note: Glass bottles have fluorinated lining.