Check Order Status
Methanol (Purge and Trap Suitable for Volatile Organic Residue Analysis), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Methanol (Purge and Trap Suitable for Volatile Organic Residue Analysis), Fisher Chemical™
Packaged under nitrogen. Submicron filtered
Have Questions?
Change view
Quantity:
1 L
Packaging:
Amber Glass
Catalog number
FGCA453-1
Price (MYR)
685.00
EA
Add to cart
Quantity:
1 L
Packaging:
Amber Glass
Request bulk or custom format
Price (MYR)
685.00
EA
Add to cart
Specifications
Chemical Name or Material
Methanol
Cleanup Symbol
Solvent
Melting Point
-98°C
Name Note
Purge and trap suitable for volatile organic residue analysis
CAS
67-56-1
View more