▸
Acids
▸
Organic Acids
▸
Acetic Acid
▸
Acetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
C2H4O2, CAS Number-64-19-7, 50mL, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C
Have Questions?
Change view
Quantity:
50 mL Poly Bottle
Catalog number
FGCA113-50
Price (MYR)
693.00
Each
Add to cart
Quantity:
50 mL Poly Bottle
Request bulk or custom format
Price (MYR)
693.00
Each
Add to cart
Specifications
pH
2.5
CAS
64-19-7
Melting Point
16°C
Boiling Point
117°C
Color
Colorless
View more
C2H4O2, CAS Number-64-19-7, 50mL, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C