Check Order Status
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Alkalimetric standard
Have Questions?
Change view
Quantity:
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number
FCMT395-100
Price (MYR)
511.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
511.00
EA
Add to cart
Specifications
Absorbance
Pass Test
Chemical Name or Material
Tris(hydroxymethyl)aminomethane
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Melting Point
168.5°C
View more