Check Order Status
Formalin Solution, 10% (Histological), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Formalin Solution, 10% (Histological), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMSF98-4
4 L
Poly Bottle
Catalog number
FCMSF98-4
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Formalin Solution
Physical Form
Liquid
Melting Point
0°C
Name Note
10%
Boiling Point
100°C
View more