▸
Biochemical Reagents
▸
Histology Reagents
▸
Formalin, Buffered, 10% (Phosphate Buffer/Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Formalin, Buffered, 10% (Phosphate Buffer/Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMSF100-4
4 L
Poly Bottle
Catalog number
FCMSF100-4
Price (MYR)
719.00
Bottle
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
719.00
Bottle
Add to cart
Chemical Identifiers
CAS
50-00-0, 67-56-1, 7558-79-4, 10049-21-5, 7732-18-5
Specifications
Chemical Name or Material
Formalin, Buffered, 10%
Physical Form
Liquid
Melting Point
0°C
Name Note
Phosphate Buffer
Boiling Point
93.9°C
View more