accumet™ Electrode Storage Solution
Actual product may vary
Fisher Chemical
accumet™ Electrode Storage Solution
Have Questions?
Catalog NumberBoiling Point
FCMSE40-1100°C
Catalog number FCMSE40-1
Price (MYR)
549.00
Each
Boiling Point:
100°C
Price (MYR)
549.00
Each
Solution of potassium hydrogen phthalate—potassium chloride conditions electrodes when not in use
Specifications
Boiling Point100°C
Capacity (English)33.8 oz.
Capacity (Metric)1000 mL
ColorClear
For Use With (Equipment)Electrode
TypeStorage Solution
Water SolubleMiscible
Unit SizeEach