▸
Solvents
▸
Hexane
▸
n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and GC/MS analysis
Have Questions?
Change view
Quantity:
4 L
Packaging:
Safe-Cote™ Amber Glass
Amber Soda Lime Glass
Catalog number
FCMH306SK4
Price (MYR)
1,051.00
Each
Add to cart
Quantity:
4 L
Packaging:
Safe-Cote™ Amber Glass
Request bulk or custom format
Price (MYR)
1,051.00
Each
Add to cart
Specifications
Chemical Name or Material
n-Hexane
Melting Point
-95°C
Absorbance
0.005 AU max. at 254nm, 0.005 AU max. at 280nm, 0.07 AU max. at 220nm, 0.25 AU max. at 210nm, 1.00 AU max. at 195nm
CAS
110-54-3
Color
Colorless
View more
Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
Tested for GC/MS suitability
Also meets ACS specifications
Intended for research use only