▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Eosin Y (Certified Biological Stain), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Eosin Y (Certified Biological Stain), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCME511-25
25 g
Glass Bottle
Catalog number
FCME511-25
Price (MYR)
483.00
Bottle
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
483.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Eosin Y
CAS
17372-87-1
ChemAlert Storage Symbol
Gray
Bio. Stain Comm. Certification
Pass Test
Dye Content
88% min.
View more