Chloroform (HPLC), Fisher Chemical™
Chloroform (HPLC), Fisher Chemical™
Chloroform (HPLC), Fisher Chemical™
Chloroform (HPLC), Fisher Chemical™
Fisher Chemical
Chloroform (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Have Questions?
Catalog NumberQuantityPackaging
FCMC60744 LGlass Bottle
Catalog number FCMC6074
Price (MYR)
925.00
Bottle
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
925.00
Bottle
Specifications
Chemical Name or MaterialChloroform
Cleanup SymbolSolvent
Melting Point-63°C
Name NoteApprox. 50ppm Pentene as Preservative
Absorbance0.01 AU max., 0.01 AU max. at 280nm, 0.05 AU max. at 270nm, 0.15 AU max. at 254nm, 0.15 AU max. at 260nm, 0.25 AU max. at 255nm, 0.32 AU max. at 250nm, 1.00 AU max. at 245nm, Pass Test
View more
HPLC mobile phase for analytical and preparative separations