Check Order Status
Citric Acid Anhydrous (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Citric Acid Anhydrous (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMA940-1
1 kg
Poly Bottle
Catalog number
FCMA940-1
Price (MYR)
552.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
552.00
EA
Add to cart
Specifications
Chemical Name or Material
Citric Acid Anhydrous
pH
1.7
CAS
77-92-9
ChemAlert Storage Symbol
Gray
Melting Point
153°C
View more