Check Order Status
Boric Acid (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Boric Acid (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMA73-500
500 g
Poly Bottle
Catalog number
FCMA73-500
Price (MYR)
484.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
484.00
EA
Add to cart
Specifications
Chemical Name or Material
Boric Acid
pH
3.8 to 4.8
Color
White
CAS
10043-35-3
ChemAlert Storage Symbol
Gray
View more