Acetic Acid, Glacial (USP/FCC/EP/BP), Fisher Chemical
Actual product may vary
Acetic Acid, Glacial (USP/FCC/EP/BP), Fisher Chemical
Acetic Acid, Glacial (USP/FCC/EP/BP), Fisher Chemical
Fisher Chemical
Acetic Acid, Glacial (USP/FCC/EP/BP), Fisher Chemical
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FCMA491-2122.5 LGlass Bottle
Catalog number FCMA491-212
Price (MYR)
1,135.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Price (MYR)
1,135.00
Each
Glass Bottle; 2.5L
Specifications
Chemical Name or MaterialAcetic Acid, Glacial
Cleanup SymbolAcid
Name Note17.4N
pH2.5
CAS64-19-7
ChemAlert Storage SymbolRed
Melting Point>14.8°C
Boiling Point117°C
ColorColorless
Health Hazard 2DANGER!
Health Hazard 3Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:2
Instability:0
DOT InformationClass 8: Corrosive
AppearanceClear and Colorless (EP/BP)
Molecular FormulaC2H4O2
Linear FormulaCH3COOH
MDL NumberMFCD00036152
UN Number2789
Synonymethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
IUPAC Nameacetic acid
Molecular Weight (g/mol)60.05
PubChem CID176
ChEBICHEBI:15366
Formula Weight60.05
Percent Purity99.0 to 100.5% (EP)
GradeBP/EP/FCC/USP
Density1.048g/cm³
Quantity2.5 L
Flash Point40°C
PackagingGlass Bottle
Physical FormLiquid
Residue after Evaporation0.01% max. (BP), 0.01% max. (EP), 0.01% max. (EP/BP)
Vapor Pressure1.52 kPa at 20°C
Unit SizeEach