▸
Solvents
▸
Isopropanols
▸
IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™
Have Questions?
Change view
Quantity:
4 L
1 L
Packaging:
Poly Bottle
Catalog number
FCMA459-4
Price (MYR)
741.00
Each
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
741.00
Each
Add to cart
Specifications
Chemical Name or Material
IPA (Isopropyl Alcohol), 70% v/v
Melting Point
-88°C
CAS
67-63-0
Color
Colorless
Health Hazard 2
WARNING!
View more