Pro-Detect™ Rapid DYKDDDDK-His Assay Kit
Actual product may vary
Fisher Chemical
Pro-Detect™ Rapid DYKDDDDK-His Assay Kit
Pro-Detect™ Rapid Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of tagged proteins in protein expression and purification systems
Have Questions?
Catalog NumberQuantity
FCMA38-50010 Assays
Catalog number FCMA38-500
Price (MYR)
1,633.00
Bottle
Quantity:
10 Assays
Price (MYR)
1,633.00
Bottle
  • Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the tag to provide a visual, color readout for detection within ten minutes.
  • Detect proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.
  • Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
  • Dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
  • Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
  • Specific—assay does not cross-react with common lysis and protein preparation reagents
  • Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
  • Long shelf life—stable for at least one year at 4°C
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detectable AnalytesDYKDDDDK-His (FLAG-His) double tagged proteins
AssayLateral Flow Rapid Assay
Detection MethodChromogenic
Product LinePro-Detect
Product TypeAssay
SpecificityProtein tag
Sufficient For10 Assays
For Use With (Application)Lateral Flow Rapid Assay
Quantity10 Assays
Shipping ConditionWet Ice
TargetProtein
Unit SizeBottle
Contents & Storage
• 10 Rapid Assay Strips
• Sample Diluent, 15 mL
Sufficient for 10 assay determinations
Store at 2–8°C.