▸
Solvents
▸
Acetonitrile
▸
Acetonitrile (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetonitrile (Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Have Questions?
Catalog Number
Quantity
Packaging
FCMA21-4
4 L
Glass Bottle
Catalog number
FCMA21-4
Price (MYR)
1,057.00
Bottle
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,057.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Acetonitrile
Cleanup Symbol
Solvent
Melting Point
-46°C
CAS
75-05-8
ChemAlert Storage Symbol
Red
View more