Precision Balances
Actual product may vary
Precision Balances
Featuring RS232 connectivity for easy communication, a backlit display and a simple interface for uncomplicated operation, these balances are perfectly designed for your workplace.
Have Questions?
Catalog NumberCapacityModelCalibrationDimensions (D x W x H)
FBS159275104200 gFPRS4202Internal317 x 201 x 93 mm
Catalog number FBS15927510
Price (MYR)
4,634.00
Each
Capacity:
4200 g
Model:
FPRS4202
Calibration:
Internal
Dimensions (D x W x H):
317 x 201 x 93 mm
Price (MYR)
4,634.00
Each
  • Standard RS-232 connectivity for easy data transfer
  • Stainless steel platform
  • Two adjustable feet that allow user to level the balance
  • Up-front level indicator - no need to look in back of balance during leveling process
  • Smart design and durable construction - small footprint which saves on benchtop space while providing a large weighing surface
  • Large, bright backlit display allows for easy viewing of results
  • Four mechanical keys enable easy operation of basic application modes
  • Weighing modes include weighing, parts counting and percent weighing
  • Includes AC adapter
  • Certifications: FCC, CE
  • Two year warranty
Specifications
CalibrationInternal
Certifications/ComplianceGLP with optional printer compliance
Connection TypeRS232
Capacity4200 g
Display TypeBacklit LCD
Readability0.01 g
IncludesAC adapter
Linearity0.02 g
Min. Sample Weight20 g
ModelFPRS4202
Repeatability0.01 g
Stabilization Time1 sec.
TypePrecision Balance
Warranty2 Years
Weighing ModesWeighing, Parts Counting, Percent Weighing
Dimensions (D x W x H)317 x 201 x 93 mm
Diameter (Metric) Outer Pan180 mm
Electrical Requirements100/240 V ∼200 mA 50/60 Hz 12/18 VA
Amperage0.5 A