Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Laboratory Surfactants and Wetting Agents
▸
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
Sodium Dodecyl Sulfate is the most commonly used detergent in protein purification and electrophoresis
Have Questions?
Change view
Quantity:
500 g
100 g
Packaging:
Poly Bottle
Catalog number
FBRBP8200-500
Price (MYR)
1,152.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
1,152.00
Each
Add to cart
Specifications
Absorbance
≤0.10 (0.1% Solution) at 260nm
Chemical Name or Material
Sodium Dodecyl Sulfate (SDS), Micropellets
Color
White
CAS
151-21-3
Physical Form
Solid
View more