Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents
Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents
Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents
Fisher Chemical
Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number FBRBP350-500
Price (MYR)
499.00
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
499.00
Each
Specifications
Chemical Name or MaterialDextrose Anhydrous
CAS50-99-7
ChemAlert Storage SymbolGray
Health Hazard 2CAUTION!
Health Hazard 3Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
View more
Dextrose is often used as a cell culture additive.