▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number
FBRBP332-500
Price (MYR)
531.00
Each
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
531.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Phosphate Dibasic Anhydrous
CAS
7558-79-4
ChemAlert Storage Symbol
Gray
Color
White
Health Hazard 1
Signal Word: CAUTION!
View more
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)
This buffer component is commonly used in tissue culture and molecular biology applications.