▸
Biochemical Reagents
▸
Carbohydrates
▸
Oligosaccharides
▸
α-Cyclodextrin (White Powder), Fisher BioReagents
Element Search
Structure Search
α-Cyclodextrin (White Powder), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP2501-25
25 g
Amber Glass
Catalog number
FBRBP2501-25
Price (MYR)
-
Quantity:
25 g
Packaging:
Amber Glass
Request bulk or custom format
Specifications
Chemical Name or Material
α-Cyclodextrin
CAS
10016-20-3
ChemAlert Storage Symbol
Gray
Melting Point
278°C
Color
White
View more
α-Cyclodextrin is used in chromatographic separations and purification methods. Useful for selective precipitation of enantiomeric, positional or structural isomers.