▸
Solvents
▸
Other Solvents
▸
Formamide
▸
Formamide (Molecular Biology), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Formamide (Molecular Biology), Fisher BioReagents
For nucleic acid hybridization and sequencing
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP227-500
500 mL
Glass Bottle
Catalog number
FBRBP227-500
Price (MYR)
1,194.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,194.00
Each
Add to cart
Specifications
Chemical Name or Material
Formamide
pH
4 to 5
Melting Point
2°C
Absorbance
0.1 max. at 280nm
CAS
75-12-7
View more
Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing. It requires pretreatment with a mixed-bed resin.