2-Mercaptoethanol (Electrophoresis), Fisher BioReagents
2-Mercaptoethanol (Electrophoresis), Fisher BioReagents
2-Mercaptoethanol (Electrophoresis), Fisher BioReagents
2-Mercaptoethanol (Electrophoresis), Fisher BioReagents
Fisher Chemical
2-Mercaptoethanol (Electrophoresis), Fisher BioReagents
Have Questions?
Catalog NumberQuantityPackaging
FBRBP176-10089.74 mLAmber Glass/Poison Pack
Catalog number FBRBP176-100
Price (MYR)
777.00
EA
Add to cart
Quantity:
89.74 mL
Packaging:
Amber Glass/Poison Pack
Request bulk or custom format
Price (MYR)
777.00
EA
Add to cart
Specifications
Alpha VectorMercaptoethanol
Chemical Name or Material2-Mercaptoethanol
CAS60-24-2
ChemAlert Storage SymbolRed
Absorbance0.05 AU max.
View more
2-Mercaptoethanol reduces disulfide linkages in proteins and peptides and is used in many enzymatic reactions as a protein stabilizer.