Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Fisher Chemical
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Have Questions?
Catalog NumberQuantityPackaging
FBRBP169-500500 gPoly Bottle
Catalog number FBRBP169-500
Price (MYR)
844.00
Each
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
844.00
Each
Specifications
Chemical Name or MaterialUrea
CAS57-13-6
ChemAlert Storage SymbolGray
Absorbance0.05 max. (5M solution) at 280nm
Melting Point131°C
View more
Urea is used as a denaturing agent for DNA and proteins.