Check Order Status
SDS-PAGE Buffer, 5X Powder, Electrophoresis, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
SDS-PAGE Buffer, 5X Powder, Electrophoresis, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP1398-92
92 g
Catalog number
FBRBP1398-92
Price (MYR)
606.00
EA
Add to cart
Quantity:
92 g
Request bulk or custom format
Price (MYR)
606.00
EA
Add to cart
Chemical Identifiers
CAS
56-40-6, 77-86-1, 151-21-3
Specifications
Chemical Name or Material
Tris-Glycine-SDS
CAS
56-40-6
ChemAlert Storage Symbol
Gray
Color
White
CAS
77-86-1
View more
Mix of tris-glycine-SDS (TGS) for electrophoresis buffer
Tris-glycine-SDS is the most commonly used running buffer for SDS-PAGE
Pre-weighed and pre-mixed ingredients to make 1L of 5X tris-glycine-SDS buffer
Eliminates the need to weigh ingredients - simply dissolve powder in pure water