DMF, Dimethylformamide, Sequencing, Fisher BioReagents
DMF, Dimethylformamide, Sequencing, Fisher BioReagents
DMF, Dimethylformamide, Sequencing, Fisher BioReagents
DMF, Dimethylformamide, Sequencing, Fisher BioReagents
Have Questions?
Catalog NumberQuantityPackaging
FBRBP1160-500500 mLGlass Bottle
Catalog number FBRBP1160-500
Price (MYR)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Dimethylformamide
pH6 to 8
Cleanup SymbolSolvent
Melting Point-61°C
AbsorbancePass Test, 1 max. at 270nm, 0.3 max. at 275nm, 0.1 max. at 295nm, 0.05 max. at 310nm, 0.01 max. (340nm to 400nm) at 340nm
View more
N,N-Dimethylformamide is commonly used in automated synthesis procedures.