Covers for Fisherbrand™ Ultrasonic Cleaners
Actual product may vary
Covers for Fisherbrand™ Ultrasonic Cleaners
Covers for Fisherbrand™ Ultrasonic Cleaners
Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
FBE1003281
also known as FBE100 3281
Ultrasonic Cleaner, 2.75 and 4.25 LLid
FBE1003285Ultrasonic Cleaner, 9.5 and 12.75 LLid
FBE1003287
also known as FBE100 3287
Ultrasonic Cleaner, 28 LLid
Catalog number FBE1003281
also known as FBE100 3281
Price (MYR)
-
For Use With (Equipment):
Ultrasonic Cleaner, 2.75 and 4.25 L
Type:
Lid
  • Easy-to-open and close cover
  • Muffles noise level
  • Bring your ultrasonic cleaner up to temperature more quickly
  • Contain and prevent spills and leakage
  • Idea for medical laboratories and industrial production processes
Specifications
TypeLid
For Use With (Equipment)Ultrasonic Cleaner, 2.75 and 4.25 L
For Use With (Application)Medical Laboratories and in Industrial Production Processes
MaterialPlastic
Unit SizeEach