Product Image
Eyela
Cone with Imhoff Settling
Fully graduated cones for determination of small amounts of settleable matter in sewage by Imhoff Sedimentation Method. Glass cone features reinforced tip for ruggedness, while the Polycarbonate cone features a leak-proof, removable closure for withdrawal of sediment for analysis. Support ring, support stand,and cone holders available and sold separately.
Have Questions?
Catalog NumberVolume (Metric)
EYE2067001000 mL
Catalog number EYE206700
Price (MYR)
7,285.00
Each
Volume (Metric):
1000 mL
Price (MYR)
7,285.00
Each
Fully graduated cones for determination of small amounts of settleable matter in sewage by Imhoff Sedimentation Method. Glass cone features reinforced tip for ruggedness, while the Polycarbonate cone features a leak-proof, removable closure for withdrawal of sediment for analysis. Support ring, support stand,and cone holders available and sold separately.
  • Glass cone features reinforced tip for ruggedness
  • Polycarbonate cone features a leak-proof, removable closure for withdrawal of sediment for analysis
  • Support ring, support stand, and cone holders available and sold separately
Specifications
DescriptionScrew Closure
Volume (Metric)1000 mL
MaterialPolycarbonate
Unit SizeEach