VertiKlean™ MAX™ Mop Heads
Actual product may vary
Corning
VertiKlean™ MAX™ Mop Heads
Clean walls, ceilings and other hard-to-reach surfaces with ease. VertiKlean™ MAX™ Mop Heads easily snap onto frames and last longer than traditional mops.
Have Questions?
Catalog NumberDimensionsSterilityMaterialHead Type
CTCVKMX027532 x 11 x 2 cmSterileRibbed Polyester Microfiber, FoamNon-sealed Edge
Catalog number CTCVKMX0275
Price (MYR)
4,519.00
Case
Dimensions:
32 x 11 x 2 cm
Sterility:
Sterile
Material:
Ribbed Polyester Microfiber, Foam
Head Type:
Non-sealed Edge
Price (MYR)
4,519.00
Case
  • Features a single layer of hydrophilic polyurethane foam laminated to textured polyester microfiber fabric
  • Backed by a semi-rigid thermoformed plate that easily snaps onto mop head frame
  • Designed for applying disinfectants to both horizontal and vertical surfaces
  • Mop heads are composed of a disposable absorbent sponge laminated to filament polyester knit fabric and backed with a thermoformed plate that easily snaps onto VertiKlean mop hardware
  • Now available with ribbed microfiber for better contamination removal
  • Pivoting mop head frame is ideal for cleaning walls, floors, and ceilings as well as hard-to-reach areas such as fume hoods, mini environments, and around light fixtures and heavy equipment
  • Sterile mop heads are validated sterile per AAMI ISO11137 guidelines
  • Autoclaving mop heads is not recommended due to thermoformed plate; however, all hardware is autoclavable
Specifications
Color CodeWhite
Dimensions32 x 11 x 2 cm
Product TypeMop Head
Head TypeNon-sealed Edge
SterilitySterile
AutoclavableNot Autoclavable
DisposableSingle-use
Color-codedNo
MaterialRibbed Polyester Microfiber, Foam
Unit SizeCase