CONSTIX™ Sealed Foam Swabs
Actual product may vary
Corning
CONSTIX™ Sealed Foam Swabs
Choose tough swabs that are ideal for cleaning. The CONSTIX™ Sealed Foam Swabs are excellent for particle removal. Use with solvents such as isopropyl alcohol.
Have Questions?
Catalog NumberLength (Metric)Handle MaterialTip StyleWidth (Metric) Tip
CTCSF-13ESD81.3 mmStat-rite ESD, Acrylic PolymerPointed0.76 mm
Catalog number CTCSF-13ESD
Price (MYR)
-
Length (Metric):
81.3 mm
Handle Material:
Stat-rite ESD, Acrylic Polymer
Tip Style:
Pointed
Width (Metric) Tip:
0.76 mm
  • Strong, resist abrasion
  • Open cell foam head for excellent cleaning and particle removal
  • Large head is ideal for larger areas
  • Excellent for applying coatings
  • Ideal for cleaning with solvents such as isopropyl alcohol
Specifications
Diameter (English) Handle0.1 in.
Length (English)3.2 in.
For Use With (Application)Ideal for cleaning with solvents such as Isopropyl Alcohol, cleaning recessed and hard to reach surfaces, applying lubricants and removing excess materials
Product TypeSwab
Diameter (Metric) Handle2.5 mm
Length (Metric)81.3 mm
Length (English) Handle2.5 in.
Length (Metric) Handle63.5 mm
MaterialFoam
Length (English) Tip0.7 in.
Length (Metric) Tip17.8 mm
Width (Metric) Tip0.76 mm
Width (English) Tip0.03 in.
AutoclavableNot Autoclavable
SterilityNon-sterile
Adhesive TypeThermal
Handle MaterialStat-rite ESD, Acrylic Polymer
Tip StylePointed
Tip MaterialPolyurethane Foam
Unit SizeCase