Amplitude™ EcoCloth™ Cleanroom Wipes
Amplitude™ EcoCloth™ Cleanroom Wipes
Actual product may vary
Amplitude™ EcoCloth™ Cleanroom Wipes
Feature enhanced sorbency and economical performance due to light weight construction. Contec™ Amplitude™ EcoCloth™ Cleanroom Wipes exceed comparable standard weight cellulose/polyester wipes in absorbency. They are ideal for the wipe down of products prior to introduction into the cleanroom.
Have Questions?
Catalog NumberDimensions (L x W)
CTCAMEC00039 x 9 in. (22.86 x 22.86 cm)
Catalog number CTCAMEC0003
Price (MYR)
-
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
  • Made of strong and durable polyester/cellulose hydroentangled nonwoven fabric
  • Economical, lightweight wiper ideal for wiping and absorbing spills in cleanroom environments; excellent for use in less critical applications
  • Exceptionally absorbent in terms of weight-to-capacity ratio
  • Low levels of particles and extractables
  • Double packaged to preserve product integrity
  • Recommended for use in ISO Class 6 and above cleanroom environments
  • General purpose cleaning in biotech, pharmaceutical, medical device, and similar industries
  • Designed for nonabrasive applications
Specifications
Product TypeCleanroom Wipe
Length (English)9 in.
Length (Metric)23 cm
Width (English)9 in.
Width (Metric)23 cm
MaterialPolyester Cellulose
AbsorbancyIntrinsic: 6.4mL/g; Extrinsic: 356mL/m²
Basis Weight57
Certifications/ComplianceISO Class 6
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)Cleanroom
Unit SizeCS