QuickConnect™ Mop Head Frames
QuickConnect™ Mop Head Frames
Actual product may vary
QuickConnect™ Mop Head Frames
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ QuickConnect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Have Questions?
Catalog NumberFor Use With (Equipment)Length (Metric)
CTC2652SSMedium and Large VertiKlean MAX Mop Heads29 cm
Catalog number CTC2652SS
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Medium and Large VertiKlean MAX Mop Heads
Length (Metric):
29 cm
  • Offer maximum flexibility for cleaning in critical environments.
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
Specifications
Color CodeWhite
Product TypeMop Head Frame
Length (English)11.5 in.
SterilityNon-sterile
Length (Metric)29 cm
AutoclavableAutoclavable
DisposableReusable
For Use With (Equipment)Medium and Large VertiKlean MAX Mop Heads
For Use With (Application)Pharmaceutical and medical device manufacturing facilities, or other similar critical or controlled environments
Color-codedNo