▸
First Aid and Medical
▸
Sharps Disposal Containers and Accessories
▸
MooreBrand™ Sharps Containers
Actual product may vary
Corning
MooreBrand™ Sharps Containers
Have Questions?
Catalog Number
Capacity (English)
CRG93268
2 gal.
Catalog number
CRG93268
Price (MYR)
3,775.00
Case
Add to cart
Capacity (English):
2 gal.
Price (MYR)
3,775.00
Case
Add to cart
Specifications
Capacity (Metric)
7.57 L
Capacity (English)
2 gal.
Color
Red
Disposable
Single-use
Stackable
Yes
Width (English)
14.5 in.
Length (English)
20.25 in.
Height (English)
18 in.