▸
Protein Analysis Reagents
▸
Bioactive Small Molecules
▸
GW 542573X
Element Search
Structure Search
Corning
GW 542573X
Selective activator of KCa2.1 channels
Have Questions?
Catalog Number
Quantity
CRG431110
10 mg
Catalog number
CRG431110
Price (MYR)
1,201.00
Pack
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
1,201.00
Pack
Add to cart
Specifications
Chemical Name or Material
GW 542573X
CAS
660846-41-3
Recommended Storage
Store at +4°C
Quantity
10 mg
Molecular Formula
C19H28N2O5
View more
Activator of small-conductance Ca2+-activated K+ channels (KCa2); selective for KCa2.1 (EC50 = 8.2 μM in HEK293 cells expressing hKCa2.1) with the profile hKCa2.1 > hKCa2.2 = hKCa2.3 > hKCa3.1 (IK).