1000 mL Vacuum Filter/Storage Bottle System
Actual product may vary
Corning
1000 mL Vacuum Filter/Storage Bottle System
Vacuum filtration systems use a square filter membrane support system that increases filtration rates with printed funnels to identify membrane type and product number. The system consists of a polystyrene filter funnel joined by an adapter ring to a removable polystyrene storage bottle with a separate sterile polyethylene cap.
Have Questions?
Catalog NumberMaterial (Membrane)Funnel CapacityPore Size
CRG431098PES1000 mL0.22 μm
Catalog number CRG431098
Price (MYR)
1,419.00
Case
Material (Membrane):
PES
Funnel Capacity:
1000 mL
Pore Size:
0.22 μm
Price (MYR)
1,419.00
Case
  • Sterile vacuum filtration system size 1L (1000 mL) with 45 mm neck size
  • Funnel contains 79 x 79 mm square membrate filter with a capacity of either 500 mL or 1000 mL
  • Receiver easily unscrews from unit and can be sealed with a plug cap for filtrate storag
  • Low liquid retention funnel design
  • Filters feature printing on the funnel for easy product identification
  • Angled hose connector simplifies vacuum line attachment
  • Receiver bottles feature easy grip sides for improved handling
  • Extra plastic storage bottles are available
  • Prefilters must be ordered separately
  • Individually packaged
  • Radiation sterilized and certified nonpyrogenic
  • Caps for receiver bottles are sterile and individually packaged
  • Units are color coded by membrane type and labeled with both membrane material and pore size
  • All membrane types are tissue culture compatible, and heat-sealed to the support grid to maximize flowrate, and reduce foaming and protein denaturization
  • Cellulose Acetate (CA) membrane for low protein binding
  • Nylon membrane for low extractables
  • Cellulose nitrate (CN) for general-purpose filtration
  • Polyethersulfone (PES) for fast flowrates and low protein binding
  • Nylon, Cellulose Nitrate and Polyetersulfone (PES) membranes are detergent-free
  • Not for parenteral use
Specifications
ConnectorsAngled Hose
DescriptionIncludes square membrane filter, graduated funnel and receiver of clear polystyrene, polyethylene neck adapter, orange HDPE plug seal
DisposableSingle-use
MaterialPolystyrene
Material (Membrane)PES
Pore Size0.22 μm
SterilitySterile
Filtration Area54.5 cm2
For Use With (Application)Filtering and storing cell culture media, biological fluids and other aqueous solutions
GraduationsYes
Quantity12/Cs.
TypeVacuum Filter System
Volume (Metric)1000 mL
Volume (Metric) Receiver1000 mL
Funnel Capacity1000 mL
For Use With (Equipment)Bottle
Unit SizeCase