Easy Grip Disposable Polystyrene Sterile Bottles
Actual product may vary
Corning
Easy Grip Disposable Polystyrene Sterile Bottles
Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
CRG430282500 mL
CRG430281250 mL
Catalog number CRG430282
Price (MYR)
571.00
Case
Capacity (Metric):
500 mL
Price (MYR)
571.00
Case
  • Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
  • Corning low profile style has easy grip sides that facilitate handling
  • Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
  • Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
  • Sterile
  • Nonpyrogenic
Specifications
Capacity (Metric)500 mL
For Use With (Application)Applications: Storage of media, buffers and other aqueous solutions
IncludesOrange HDPE plug seal
MaterialPolystyrene
No. per Case24/Cs.
No. per Pack2/Pk.
Quantity24 Case
SterilitySterile
Mouth4.5 cm
DisposableSingle-use
Product TypeGraduated Easy Grip Storage Bottle
Unit SizeCase