Pure Ethanol products are taxed according to the government schedule of excise taxes detailed in US CFR Part 27 (211.1119). This excise tax is included in the cost of this product. There is no need for Federal Permits
Have Questions?
Catalog Number
Quantity
Packaging
CRG3916
24 x 0.125 gal.
Glass Bottle
Catalog number CRG3916
Price (MYR)
2,838.00
Case
Quantity:
24 x 0.125 gal.
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,838.00
Case
Specifications
Chemical Name or MaterialEthanol
Melting Point-114°C
Name Note200 Proof (100%)
CAS64-17-5
ColorClear
View more
For the extraction and precipitation of DNA and RNA, can be used full strength or diluted with endonuclease-free water to desired dilution.