96-Well Clear Bottom Black or White Polystyrene Microplates
Actual product may vary
Corning
96-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates permit direct microscopic viewing. For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays.
Have Questions?
Catalog NumberColor
CRG3632White, Clear Bottom
Catalog number CRG3632
Price (MYR)
4,333.00
Case
Color:
White, Clear Bottom
Price (MYR)
4,333.00
Case
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Specifications
    FormatStandard
    LidWithout Lid
    Array8 x 12
    ColorWhite, Clear Bottom
    MaterialPolystyrene
    No. of Wells96
    Volume (Metric) Working75 to 200 μL
    SterilityNon-sterile
    Surface TreatmentNon-treated
    Well ShapeFlat Bottom (F-well)
    For Use With (Application)Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    No. per Case100/Cs.
    No. per Pack25/Pk.
    Quantity100/Cs.
    Unit SizeCase