CellBIND™ Surface Cell Culture Flasks
Actual product may vary
Corning
CellBIND™ Surface Cell Culture Flasks
Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.
Have Questions?
Catalog NumberCapacity (Metric)Neck StyleNo. per Case
CRG3292250 mLAngled50/cs.
Catalog number CRG3292
Price (MYR)
3,764.00
Case
Capacity (Metric):
250 mL
Neck Style:
Angled
No. per Case:
50/cs.
Price (MYR)
3,764.00
Case
  • 25 to 225 cm2 available growth area
  • Manufactured from optically clear virgin polystyrene
  • Treated with a Corning™ CellBIND™ surface treatment for enhanced cell attachment
  • Printed with lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation
  • Nonpyrogenic
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
BarcodeNo
Capacity (English)8.45 oz.
Closure ColorOrange
Closure MaterialHDPE
Capacity (Metric)250 mL
MaterialPolystyrene
For Use With (Application)For optimal cell attachment
IncludesLot numbers for ease in traceability
Neck StyleAngled
No. per Case50/cs.
ShapeRectangular
Surface CoatingCorning Cellbind
GraduatedYes
SterilitySterile
Bottom ShapeU-shape
Closure TypeVented
Culture Area175 cm²
Closure Size33 mm
Unit SizeCase